Learn More
Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 680, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen A21102
Description
This secondary antibody is designed for fluorescent Western blot detection on various near-infrared fluorescence instruments. This antibody can be used for multi-color and multiplexing detection when using other antibodies conjugated to compatible Alexa Fluor™ dyes and wavelengths. Other applications of this antibody include immunofluorescent and fluorescent imaging applications when using instrumentation with appropriate excitation and detection capabilities. Product will be shipped at Room Temperature.
Sheep IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Sheep IgG (H+L) Cross-Adsorbed in Ovine samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Sheep IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 680 | |
Donkey | |
purified | |
RUO | |
Ovine | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.