missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Donovan S3 Shoe

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.

£118.00 - £120.00

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Brown
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
View More Specs

Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17532646
View Documents
Emma Safety Footwear
MM99456939
39
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17542646
View Documents
Emma Safety Footwear
MM99456940
40 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17552646
View Documents
Emma Safety Footwear
MM99456941
41
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17562646
View Documents
Emma Safety Footwear
MM99456942
42
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17572646
View Documents
Emma Safety Footwear
MM99456943
43
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17582646
View Documents
Emma Safety Footwear
MM99456944
44
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17592646
View Documents
Emma Safety Footwear
MM99456945
45
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17502656
View Documents
Emma Safety Footwear
MM99456946
46
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17512656
View Documents
Emma Safety Footwear
MM99456947
47
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17522656
View Documents
Emma Safety Footwear
MM99456948
48
£120.00
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications

Specifications

ISO 20345:2011
Brown
Hydro-Tec Antibacterial
Extra-wide Width
Safety Shoe
Laces
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Nubuck, Oiled Nubuck
High
Videos
SDS
Documents

Documents

Product Certifications