Learn More
BINDER™ Door Gasket
This Door Gasket is made of FKM. For use with assorted BINDER™ Drying and Heating Chambers.
Brand: BINDER™ 8012-1833
Additional Details : Weight : 0.00100kg
Description
- Made of FKM
- Silicone-free
Specifications
Door gasket - made of FKM, silicone-free | |
BINDER Chambers FD 720 Drying and Heating Chambers with Forced Convection | |
Door Gasket | |
FKM |