Learn More
Miele Professional™ Dosing system
Dispenser module DOS K 60 for 5-10 litre canisters with level indicator for the automatic dispensing of liquid cleaning agents (G 7831)
Brand:  Miele Professional™ 6940330
Product Code. 10610724
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Dispenser monitoring – in accordance with DIN EN ISO 15883
  • For dispensing liquid cleaning agent/chemical disinfectant
  • 300 mm siphon for 5-10 litre containers
  • Conversion kit for siphons for 10-30 l canisters available
  • Dispenses up to 120 ml/60 seconds
Specifications
Specifications
Dispensing System
DOS K 60/1 Dispensing Module
Washer-disinfectors, laboratory
1 Each