Learn More
Socorex™ Dosys™ Premium Two-ring Automatic Syringe
Dose with excellent reproducibility using a corrosion-resistant two-ring handle automatic syringe. Socorex™ Dosys™ Premium Two-ring Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
Brand:  Socorex™ 164.0505
Code : 03
Additional Details : Weight : 0.32040kg
 View more versions of this product
Product Code. 10111542
  • £214.00 / Each
Estimated Shipment: 05-11-2024
to see stock.
Description
Description
  • Two-ring handle.
  • Self-refilling automatic syringe.
  • Detachable Luer lock valve system.
  • High chemical resistance.
  • Corrosion resistant.
  • Optimal hand balance and ergonomics.
  • Fits either hand.
  • Requires limited effort.
  • Fast and reliable volume setting.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness without O-ring.
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
  • Autoclavable (except for protective PVC sleeve).
Specifications
Specifications
Autoclavable
Syringe, 1M silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spares for valve, operating instructions.
0.5 mL
1/ea.
0.5 to 5 mL
Yes
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Automatic Syringe, Two-ring
Dosys™ Premium 164
<0.5% to <0.2%
Luer Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products