missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Dot Blot Hybridisation Manifold System
Click to view available options
Device Holds:
48 Samples
96 Samples
Unit Size:
Each
Description
- High density acrylic
- Microfiltration manifold
- Allows unfractionated samples to be immobilised onto membrane
- Immediate screening with nucleic acid probes or antibodies
- Upper manifold - allows sample to be loaded into each dot/slot
- Stainless steel thumbscrews and precision-lapped mating surfaces
- Template to allow membrane to be cut down to size
- Vacuum Port, can be connected to water aspirator or vacuum pump
- Vacuum reservoir to serve as collection chamber for filtrate
- Requires vacuum pump or water aspirator equivalent to 600mm Hg (0.8 bar)
- DHM-48: Used for 5 x 12.8cm membrane - 48 sample throughput
- DHM-96: Used for 7.5 x 11.3cm membrane - 96 sample throughput
- Compatible with multichannel pipettes and standard 96-well microtitre plates
Specifications
Specifications
| Device Holds | 96 Samples |
| Type | Dot Blot Hybridization System |
| Includes | 96-well Dot Blot Hybridisation Manifold in 8 x 12 array,comprising paired upper dot-forming modules for dots3mm in diameter, and lower vacuum chamber module |
| No. of Slots | 96 |
| Temperature (Metric) Operating | 65°C |
| Volume (Metric) Sample | 100 μL |
| Dimensions (L x W x H) | D x W x H: 7.5 x 18.5 x 7 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction