Learn More
Sampling Systems™ Double Bagged - USP Class VI Spoons
BIOPHARMA
Double Bagged - USP Class VI Spoons are made from high grade medical USP class VI HDPE. They are available in 3 mL and 10 mL. This premium range is double bagged. A box liner is also used giving a third layer of protection.
Brand:  Sampling Systems™ 9088H-10S
 View more versions of this product
Product Code. 17584696
  • £111.00 / Pack of 50
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These sterile spoons are commonly used in the manufacture and sampling of cosmetics, food and pharmaceuticals.
  • The spoons are designed to be used straight from the bag.
  • The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The funnels are sterilised by gamma irradiation.
  • USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • Cleanroom Manufactured - The spoons are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each spoon is 100% inspected before packing.
Specifications
Specifications
HDPE
10 mL
Sterile
Double Bagged