Learn More
R&L Enterprises™ Double Burette Clamp
Securely holds burettes in place during the construction of laboratory retort stands. R&L Enterprises™ Double Burette Clamps are suitable for burettes up to 100mL capacity.
Brand:  R&L Enterprises™ 3596
Product Code. 11749027
  • £70.65 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Diecast
  • Without rod
  • With a single plastic headed thumbscrew to secure the clamp to retort rods, vertically or horizontally
Specifications
Specifications
1 Ea.
Double Clamp
with a single Plastic headed thumbscrew to secure the clamp to retort rods
For use up to 100mL burette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Double Burette Clamp > For Use With: For use up to 100mL burette