Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
£1220.00 - £2355.00
Specifications
Includes | Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware |
---|---|
Product Type | Mobile Unit Kit |
Product Code | Brand | Material | Width (English) | Width (English) Shelf | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Width (English) | Width (English) Shelf | Price | Quantity & Availability | |||
15906991
|
Metro™
LBTM21S |
Stainless Steel | 21 in. | 21 in. |
£2320.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15936991
|
Metro™
LBTM24S |
Stainless Steel | 24 in. | 24 in. |
£2355.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15946991
|
Metro™
LBTM24X3 |
Polymer | 24 in. | 24 in. |
£2095.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15976981
|
Metro™
LBTM18S |
Stainless Steel | 18 in. | 18 in. |
£2280.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15916991
|
Metro™
LBTM24C |
Steel | 24 in. | 24 in. |
£1280.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15986981
|
Metro™
LBTM18X3 |
Polymer | 18 in. | 18 in. |
£2015.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15926991
|
Metro™
LBTM24K3 |
Steel | 24 in. | 24 in. |
£1930.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15996981
|
Metro™
LBTM21C |
Steel | 21 in. | 21 in. |
£1250.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966981
|
Metro™
LBTM18K3 |
Steel | 18 in. | 18 in. |
£1835.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15956981
|
Metro™
LBTM18C |
Steel | 18 in. | 18 in. |
£1220.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
- Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware | |
Mobile Unit Kit |