missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Double Diffusion Plates
Microgen Bioproducts double Diffusion Plates for use with the specific fungal serology kits (FSK1,FSK3 and FSK4)
Brand: Novacyt - Microgen Bioproducts FSKDDP
Additional Details : Weight : 0.50000kg
Description
- Double diffusion plates.
- These plates are specially formulated to optimise the detection of precipitating antibodies.
Specifications
Not Autoclavable | |
325 μL | |
Sterile | |
Multi-Well Plate | |
4 mm wells, 8 mm wells, 12 mm wells |
Flat | |
Rectangular | |
Physical Surface Treatment | |
2 mm |