missing translation for 'onlineSavingsMsg'
Learn More

Water, Double Distilled, Pyrogen-Free, Chem-Lab

Suitable for laboratory use. Water, Double Distilled, Pyrogen-Free, Chem-Lab has been ultra-purified to remove bacteria, organic and inorganic particles, viruses, minerals, salts and metals, at LF<5μS. Packaged in a sterilized container to maintain purity.

£44.85 - £82.30

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11983084
View Documents
Chem Lab
CL02.0208.5000
5 L
£44.85
5 L
In Stock
to see stock.
Edge
Add to basket
 
11963345
View Documents Promotion Details
Chem Lab
CL02.0208.9010
10 L
£82.30
10 L
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Distilled water is produced by boiling water and then condensing the vapor into a clean container, leaving solid contaminants behind. Double-distilled water produces an even higher level of purification. It is prepared by slow boiling the uncontaminated condensed water vapor from a prior slow boiling.
Specifications

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
0°C
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.016g/mol
Double-Distilled
Liquid
Colorless
100°C
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
LF < 5μS
1.00g/mL
Videos
SDS
Documents

Documents

Product Certifications