Learn More
BRAND™ Double-Ended PTFE Magnetic Stirring Bars
Excellent centering
Brand:  BRAND™ 137935
216.84 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 1.86000kg
 View more versions of this product
Product Code. 10668881
  • £271.42 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Small contact surface
  • High turbulence
  • Works well even at low speeds
  • Specifications
    Specifications
    8 mm
    10
    54 mm
    Dumbbell