Learn More
Hughes Double Nozzle Handheld Eye/Face/Body Wash
Product Code. 17782338
missing translation for 'orderingAttributeHoverText'
Color:
Red
missing translation for 'unitSize'
Each
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 17782338
missing translation for 'mfr': Hughes HEWS5022RB
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
A wall mounted, handheld eye, face and body wash with two nozzles. Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances.
  • A wall mounted, handheld eye, face and body wash with two nozzles
  • Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances
  • 5 m of reinforced hose makes these handheld units particularly useful for treating disabled personnel
  • The lock-on actuating button for ease of use meets European and international standards
  • Fitted with dust covers, a non-return valve and a flow regulator
Specifica
Color Red
Height (Metric) 20 cm
Product Type Eye, Face and Body Combination Wash Stations
Width (Metric) 18 cm
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt