Learn More
VELP Scientifica™ Double Rod Mini H-Stand Set
The double rod mini H-stand set for disperser tools is designed for light weight instruments and comes complete. This stand is designed to avoid rotation of attachments and instruments.
Brand:  VELP Scientifica™ A00000383
Additional Details : Weight : 1.00000kg
Product Code. 17110206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
DESCRIPTION
  • Designed for light weight instruments and comes complete with all parts
  • Designed to avoid rotation of attachment and instrument through the use of the double rod and bosshead clamp
Specifications
Specifications
Stand
300 x 430 mm
Not Autoclavable