missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Double wall carton, 229mm x 152mm x 152mm
Brand: Manutan A329293
This item is not returnable.
View return policy
Description
Greatly improved strength and rigidity over single wall boxes the double wall gives maximum protection for fragile products and supports heavier items. Ideal for export.- Internal length: 229mm
- Internal height: 152mm
- Load capacity: 5kg
Specifications
229 x 152 x 152 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction