missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Double wall carton, 508mm x 508mm x 508mm
Brand: Manutan A330127
This item is not returnable.
View return policy
Description
Greatly improved strength and rigidity over single wall boxes the double wall gives maximum protection for fragile products and supports heavier items. Ideal for export.- Internal length: 508mm
- Internal height: 508mm
- Load capacity: 25kg