missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Double Wrap Tryptic Soy Broth Media Bottle
Terminally sterilized inside of autoclavable double-bags
Brand: BD 257213
Additional Details : Weight : 0.80000kg
Description
- Free from environmental contaminants and particulate matter
Specifications
ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP | |
Liquid | |
Microbiology Media | |
Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens | |
100 mL |
5°C to 25°C | |
Tryptic Soy Broth | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L | |
Bottle |