Learn More
Gilac™ Dough Container
Dough Container is ideal for dough, pastries, chocolate molds storage, with rounded corners and bottom for easy cleaning.
£11.46 - £15.45
Product Code | Brand | Volume (Metric) | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||
17988491
|
Gilac™
G179431 |
9 L | 600 x 400 x 55 mm | White |
£11.46
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17958491
|
Gilac™
G179410 |
14 L | 600 x 400 x 76 mm | Blue |
£13.12
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17928401
|
Gilac™
G179409 |
14 L | 600 x 400 x 76 mm | Red |
£13.12
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17958401
|
Gilac™
G179441 |
14 L | 600 x 400 x 76 mm | White |
£12.28
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17998391
|
Gilac™
G179418 |
14 L | 600 x 400 x 76 mm | Yellow |
£13.12
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17938561
|
Gilac™
G179428 |
15 L | 600 x 400 x 90 mm | Blue |
£13.42
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17928611
|
Gilac™
G179411 |
15 L | 600 x 400 x 90 mm | Green |
£13.42
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17938811
|
Gilac™
G179477 |
15 L | 600 x 400 x 90 mm | Gray |
£13.42
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17978611
|
Gilac™
G179434 |
15 L | 600 x 400 x 90 mm | Red |
£13.42
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17938611
|
Gilac™
G179423 |
15 L | 600 x 400 x 90 mm | Yellow |
£13.42
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17948401
|
Gilac™
G179421 |
15 L | 600 x 400 x 90 mm | White |
£12.20
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17928561
|
Gilac™
G179420 |
15 L | 600 x 400 x 90 mm | Translucent |
£12.64
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17958701
|
Gilac™
G179430 |
23 L | 600 x 400 x 130 mm | White |
£15.45
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- 600 x 400 dough container, ideal for dough, pastries, chocolate molds... storage.
- Stacking with and without lid, maximum stacking height capacity.
- Innovative design for even greater convenience:
- Rounded corners and bottom for easy cleaning.
- Vertical sides without inner edge avoids marking the dough.
- The entire surface of the container is usable.
- Completely flat bottom means dough detaches easily, no loss, no food deposit left.
- Optimised side reinforcements allows you to customise containers with your logo.
- Rounded outside corners guarantee a damage-free working environment.
- Translucent container facilitates the control of the contents.
- Allows you to monitor dough rising without lifting the container.
Specifications
Dough Container | |
Polyethylene |