missing translation for 'onlineSavingsMsg'
Learn More
Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals Product Code.: 11369075

Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals

Product Code. 11369075
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11369075

Brand: Thermo Scientific Alfa Aesar L14257.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dowex™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
SMILES *

Specifications

Odor Amine-like
Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 1X8 100-200 (Cl)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462547 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14257.30,L14257.18,L14257.0B
Lot NumberA0460190 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14257.30,L14257.18,L14257.0B
Lot NumberZ28J006 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog NumberL14257.30,L14257.18,L14257.0B
Lot Number10224593 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberL14257.30,L14257.18,L14257.0B
Lot Number10244247 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog NumberL14257.30,L14257.18,L14257.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.