missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 50WX8-400 ion-exchange resin, Thermo Scientific Chemicals

£63.20 - £811.00

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10143821
View Documents
Thermo Scientific Acros
335351000
100 g Plastic Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10431022
View Documents
Thermo Scientific Acros
335355000
500 g Plastic Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10454415
View Documents
Thermo Scientific Acros
335350025
2.5 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

11119-67-8
MFCD00132726
AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Beads
15% max. (on 100 mesh, cumulative), 15% max. (through 325 mesh)
MFCD00132726
Beige to Brown
AmberChrom™ 50WX8 Ion Exchange Resin
11119-67-8
wet volume capacity, H form min. (meq/mL): 1.7
H form: 50 to 58%
Solubility in water: insoluble.
Dowex™ 50WX8
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Dowex 50WX8

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only