missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxorubicin Hydrochloride, Fisher BioReagents
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Packaging:
Amber Glass
Description
Doxorubicin Hydrochloride
Doxorubicin HCL is a chemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. It blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis.

Specifications
Specifications
Decomposition Information | 216°C |
Melting Point | 216°C |
Color | Orange |
Assay Percent Range | (by HPLC): ≥95% |
Packaging | Amber Glass |
Quantity | 1 mg |
Merck Index | 15, 3487 |
Solubility Information | Soluble in water |
Formula Weight | 580 |
Physical Form | Powder or Solid |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling



Product Identifier
- Doxorubicin hydrochloride
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Doxorubicin Hydrochloride, Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction