missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin Hydrochloride, Fisher BioReagents

Product Code. 10767945
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Doxorubicin Hydrochloride

Doxorubicin HCL is a chemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. It blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name (7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Decomposition Information 216°C
Melting Point 216°C
Color Orange
Assay Percent Range (by HPLC): ≥95%
Packaging Amber Glass
Quantity 50 mg
Merck Index 15, 3487
Solubility Information Soluble in water
Formula Weight 580
Physical Form Powder or Solid
Chemical Name or Material Doxorubicin Hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Doxorubicin hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'