missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxorubicin hydrochloride, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 579.99 |
Chemical Name or Material | Doxorubicin hydrochloride |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling


Product Identifier
- Doxorubicin Hydrochloride
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- H302-Harmful if swallowed.
- H350-May cause cancer.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Doxorubicin Hydrochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Doxorubicin hydrochloride, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction