missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline (hyclate), MedChemExpress
MedChemExpress Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity.
Brand: Medchem Express HY-N0565B
This item is not returnable.
View return policy
Description
Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity.Specifications
Doxycycline (hyclate) | |
Yellow | |
C22H24N2O8.1/2C2H6O.ClH.1/2H2O | |
Doxycycline hydrochloride hemiethanolate hemihydrate WC2031 | |
O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])[C@@H](O)[C@]3([H])[C@@H](C)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N.Cl.[0.5H2O].[0.5C2H6O] | |
512.94 | |
Research | |
COVID-19-immunoregulation |
24390-14-5 | |
Research | |
1 g | |
DMSO : 240 mg/mL (467.89 mM; Need ultrasonic) ∣H2O : 125 mg/mL (243.69 mM; Need ultrasonic) | |
512.94 | |
98.0% | |
Powder |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction