missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DPBS (10X), calcium, magnesium

DPBS (10X), calcium, magnesium
£21.60 - £189.00
Specifications
Additive | No Sodium Pyruvate |
---|---|
Capacity | 500 mL |
Dilution | 10 X |
Product Type | DPBS |
For Use With (Application) | Mammalian Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11560466
![]() |
Gibco™
14080089 |
10 x 500 mL |
£189.00
5000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11550466
![]() |
Gibco™
14080048 |
500mL |
£21.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility (14080-055). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications
No Sodium Pyruvate | |
10 X | |
Mammalian Cell Culture | |
4.3 to 4.7 | |
Liquid |
500 mL | |
DPBS | |
2700 to 3200 mOsm/kg | |
For in-vitro diagnostic use |
Safety and Handling
missing translation for 'shelfLife' : 24 Month(s)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title