missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS (10X), no calcium, no magnesium
GREENER_CHOICE

Product Code. p-7075823
Click to view available options
Quantity:
100mL
500mL
10 x 500 mL
20 x 100 mL
Unit Size:
100mL
2000mL
5000mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11520486

Brand: Gibco™ 14200059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

DPBS (10X), no calcium, no magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 6 to 7
Physical Form Liquid
Quantity 100mL
Includes No Sodium Pyruvate
Osmolality 2500 to 2900 mOsm/kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.