missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, calcium, magnesium

DPBS, calcium, magnesium

Brand:  Gibco™ 14040224

Code : 50

Additional Details : Weight : 1.05000kg

 View more versions of this product

Product Code. 10572703

  • £128.00 / 6 L
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications

Specifications

No Sodium Pyruvate
2° to 8°C
DPBS
Mammalian Cell Culture
7 to 7.2
Liquid
1000 mL
1 X
6 x 1000 mL
270 to 310m Osm/kg
For in-vitro diagnostic use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers