missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DPBS, no calcium, no magnesium

DPBS, no calcium, no magnesium
Brand: Gibco™ 14190169
80.29 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.Universal Bag Packaging
Gibco™ universal bags offer flexible packaging that allows you to integrate a single-use, disposable large volume media container with a wide variety of automated cell culture instruments, disposable bioreactor systems, and sterile connection processes, minimizing the need for costly customization.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Specifications
No Sodium Pyruvate | |
1 X | |
10 x 500 mL | |
270 to 300 mOsm/kg | |
Liquid |
15°to 30°C | |
DPBS | |
Mammalian Cell Culture | |
7 to 7.3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ DPBS, no calcium, no magnesium > 10 x 500mL
Spot an opportunity for improvement?Share a Content Correction