missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPCPX, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Potent and selective A1 adenosine receptor antagonist, both in vitro and in vivo. Ki values are 3.9, 130, 50 and 4000 nM for human A1, A2A, A2B and A3 receptors respectively.
Specifications
Specifications
CAS | 102146-07-6 |
Molecular Formula | C16H24N4O2 |
MDL Number | MFCD00055117 |
Quantity | 100 mg |
Synonym | 8-cyclopentyl-1,3-dipropylxanthine, dpcpx, 8-cyclopentyl-1,3-dipropyl-1h-purine-2,6 3h,7h-dione, 1,3-dpcpx, 1,3-dipropyl-8-cyclopentylxanthine, gnf-pf-2224, dipropylcyclopentylxanthine, unii-9ptp4foi9e, 9ptp4foi9e, chembl183 |
InChI Key | FFBDFADSZUINTG-UHFFFAOYSA-N |
SMILES | CCCN1C2=C(NC(=N2)C2CCCC2)C(=O)N(CCC)C1=O |
IUPAC Name | 8-cyclopentyl-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione |
Molecular Weight (g/mol) | 304.39 |
PubChem CID | 1329 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'