missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DpnI (10 U/μL)

Product Code. p-4521639 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,000 units
2,500 units
500 U
Unit Size:
1000 units
2500 units
500 units
This item is not returnable. View return policy

Product Code. 10669500

Brand: Thermo Scientific™ ER1701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at Gm6A^TC sites with DpnI restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: MalI).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Applications

  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP

Note

DpnI requires the presence of N6-methyladenine within the recognition sequence to cleave DNA. DNA purified from a dam+ strain will be a substrate for DpnI. DpnI will only cleave fully-adenomethylated dam sites. Hemi-adenomethylated dam sites DpnI cleaves 60X more slowly. DpnI, Bsp143I, and MboI all recognize the same sequence, but have different methylation sensitivities and cleavage sites. Assayed using pBR322 DNA.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Incubator Temperature 37°C
Quantity 500 U
Product Type DpnI

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.