Learn More
Hach Lange™ DR 3900 Spectrophotometer with RFID technology
With RFID for easy method update, sample ID and Certificate of Analysis.
Brand: Hach Lange™ LPV440.99.00001
This item is not returnable.
View return policy
Description
- Sample preparation: reduced handling steps, precise dosing
- Sample analysis: automatic test recognition, expiration date check etc.
- Documentation of results: versatile data management, control cards
- With your daily routine of water analysis in mind the DR3900 spectrophotometer is optimised for safe processes and meaningful reports
- Control-parameters like Ammonium, COD, Phosphate, Nitrogen and many others are easy to perform
- The handling of tests and photometer is well designed to avoid any errors in your water analysis
Specifications
Spectrophotometer for water analysis | |
DR3900 | |
Reference beam, spectral |
Product Content Correction
Product Title
Hach Lange™ DR 3900 Spectrophotometer with RFID technology > Height: 255mm
Spot an opportunity for improvement?Share a Content Correction