Learn More
Dräger™ 7000 Tube Opener
An accessory for detector tube systems.
Brand:  Dräger™ 6401200
Product Code. 10200383
  • £211.00 / Each
Explore more special offers
Description
Description
  • Accessory for detector tube systems
  • For scoring and breaking tips off individual Draeger detector tubes
  • Glass tips safely stored for proper disposal
  • Aids in breaking tube ampoules
Specifications
Specifications
Detector Tube Systems
7000 Tube Opener
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Dräger™ 7000 Tube Opener > 7000 Tube Opener