Learn More
Dräger™ CMS Analyzer with Remote Sample Pump
Product Code. 10468413
Click to view available options
Type:
CMS Analyzer
Unit Size:
Each
Product Code. 10468413
Brand: Dräger™ 8317700
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easy spot measurements in three steps: insert chip, start measurement, read measurement result on the LCD display
  • Remort-system allows access to difficult places with additional pump and extension hose
  • Additional advantage of Draget-Tubes
  • Unaffected with type of gases measured
  • Single button operations
  • Multilaungual displays
  • Audible signal sounds after each operating step
  • Replaceable batteries provides backup of seven hours measurement
Certifications
Cenelec , UL, UL/CSA
Specifications
Type CMS Analyzer