Learn More
Dräger™ CMS Chips: Perchloroethylene
Suitable for 10 measurements of one gas
Brand: Dräger™ 6406040
This item is not returnable.
View return policy
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
5 to 30mg/L | |
40 to 330 sec. | |
15° to 40°C |
Perchloroethylene CMS Chip | |
5 to 500 | |
59°F to 104°F |