missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Carbon Monoxide

Product Code. p-6467003
Click to view available options
Measurement Range:
0.3 to 7%
10 to 3000
1:2.5 to 150
2 to 50 ppm
5 to 700
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12806673

Brand: Dräger™ CH29901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Carbon Monoxide
Humidity Range 50mg/L
Measurement Range 0.3 to 7%
No. of Strokes 1
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 122°F
Temperature Range (Metric) 0°C to 50°C
Tube Type Carbon monoxide 0.3%/b
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.