missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Carbon Dioxide

Detector tubes

Brand:  Dräger™ CH20301

Code : 50

Additional Details : Weight : 0.15228kg

 View more versions of this product

Product Code. 10113112

  • £129.00 / Pack of 10
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Specifications

Carbon Dioxide
5 to 60%
10
32°F to 104°F
Carbon dioxide 5%/A
50mg/L
1
2 min.
0°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers