Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrochloric Acid
For making spot measurments for various gases and vapors
Brand: Dräger™ 6728181
This item is not returnable.
View return policy
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Hydrochloric Acid 50/a | |
50 to 5000ppm | |
10 | |
50°F to 122°F |
15mg/L | |
1/10 | |
5 min./30 sec. | |
10° to 50°C |