Learn More
Dräger™ Short-Term Detector Tubes - De Tube Monostyrene
Brand: Dräger™ 6723301
This item is not returnable.
View return policy
Description
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Monostyrene 10/a | |
10 to 200ppm | |
10 | |
59°F to 95°F |
Less than15mg/L | |
15 | |
3 min. | |
15° to 35°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
- drager tubes styrene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.