Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Brand:  Dräger™ 6728041
Code : 50
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10142032
  • £168.22 / Pack of 10
Estimated Shipment: 15-07-2024
to see stock.
Specifications
Hydrogen Sulfide 0.5/a
0.5 to 15ppm / 5 to 150ppm
10
32°F to 104°F
3 to 30mg/L
10
6 min.
0°C to 40°C