Learn More
Dräger™ Short-Term Detector Tubes - Ozone
Product Code. p-4909569
Click to view available options
Components Measured:
Ozone 0.05/b
Ozone 10/a
Measurement Range:
0.05 to 0.7ppm
20 to 300ppm
Unit Size:
Pack of 10
Product Code. 10120921
Brand: Dräger™ 6733181
For making spot measurements for various gases and vapors
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Ozone 0.05/b
Measurement Range 0.05 to 0.7ppm
Quantity 10/Box