Learn More
Dräger™ Short-Term Detector Tubes - Phosgene
Brand:  Dräger™ 8101521
 View more versions of this product
Product Code. 11368291
  • £94.05 / Each
Explore more special offers
Description
Description
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Specifications
Phosgene 0.02/a
0.02 to 1ppm
10
32°F to 104°F
3 to 15mg/L
20/40
6 to 12 min.
0°C to 40°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • DR PHOSGEN 0,02/A - 8101521
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contains : phosgene