Learn More
Dräger™ X-plore™ 5500 PC Full Face Mask
The Dräger™ X-plore™ 5500 EPDM full face mask is the right solution for environments that require increased respiratory protection and a clear field of vision
Brand: Dräger™ R55270
Code : Z1
Additional Details : Weight : 0.95400kg
Description
The double-layer face seal with triple sealing action ensures reliable protection and a secure, leak-free seal regardless of your face shape
- Double-layer face seal with triple sealing action
- Bayonet connection for easy and secure filter attachment. Both filters are inserted in a top-down direction
- The swept-back low profile design ensures an optimal field of vision and even weight distribution with superb freedom of movement
- Impact-proof polycarbonate material (with plastic frame) lens provides excellent, wide field of vision and extreme chemical, thermal, and mechanical resistance
- 5-point harness that allows quick and easy donning without hair entanglement
- Three sizes in one, fits most face and head sizes
- EPDM mask body with high ageing resistance
- Two side-positioned bayonet connections for use with the Dräger™ X-plore™ Twinfilter cartridge series
Certifications
CE Marked
Compliance
EN136 Cl.2 and NIOSH
Specifications
NIOSH, CE, EN136 Cl.2 | |
Full Face Mask | |
Right solution for environments that require increased respiratory protection and a clear field of vision |
EPDM, Polycarbonate Lens | |
Universal | |
4-point |