Learn More
Dräger™ Short-Term Detector Tubes: Ammonia
Detector tubes
Brand: Dräger™ 6733231
Code : 50
Additional Details : Weight : 0.06956kg
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Detector Tubes | |
Less than20mg/L | |
5 | |
1 min. | |
10° to 50°C |
Ammonia | |
2 to 30 | |
10 | |
50°F to 122°F | |
Ammonia 2/a |
Safety and Handling
- AMMONIA 2/A
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Informations supplémentaires
- MIXTURE LIST-Contains : ammonia