Learn More
Kern Sohn Draft Shield Rear Panel
Draft shield rear panel with integrated ioniser to neutralise electrostatic charge.
£540.03 - £1733.32
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17351513
|
Kern Sohn
ABP-A01 |
ABP Range Balances |
£1733.32
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17371553
|
Kern Sohn
ALJ-A03 |
ALJ Range Balances |
£540.03
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Fitted in place of the existing glass rear panel of the draft shield
- Includes rear panel, ionizer, and power supply
Specifications
Plastic | |
Rear Panel, Ionizer, and Power Supply |