Learn More
Invitrogen™ DRAQ7™ Dye
DRAQ7 Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry.
£133.00 - £339.00
Product Code | Brand | Quantity | Content And Storage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Content And Storage | Price | Quantity & Availability | ||||
16327895
|
Invitrogen™
D15106 |
200 Assays | Store at room temperature. |
£339.00
Pack of 200 |
In Stock
Log in to see stock. |
||||
16317895
|
Invitrogen™
D15105 |
50 Assays | Store at room temperature. |
£133.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
Description
This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.
• Stable at room temperature
• Rapid staining of dead cells without wash steps
• No UV excitation and no emission spectral overlap with PE
DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.
Specifications
Store at room temperature. | |
Dye |