missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Buffers (10X)

Product Code. p-200086898 Shop All Thermo Scientific Products
Click to view available options
Color:
Colorless
Green
Unit Size:
Pack of 10
Pack of 4
This item is not returnable. View return policy

Product Code. 11360600

Brand: Thermo Scientific™ B71

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific 10X DreamTaq Buffer is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase.

Thermo Scientific DreamTaq Buffer (10X), available in colorless and green formats, is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase. It contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2

In addition, 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes. The density reagent allows direct loading of PCR products on a gel. The blue dye (migrates with 3–5 kb DNA fragments in 1% agarose gel) and the yellow dye (migrates faster than 10 bp DNA fragments in 1% agarose gel) are included for monitoring electrophoresis progress. The dyes have excitation peaks at 424 nm and 615 nm.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless DreamTaq Buffer.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DreamTaq Green Buffer
Color Green
Concentration 10X
For Use With (Application) Hot-start PCR
Quantity 4 x 1.25 mL
Formulation Nonidet P40 Ethyl phenolpoly(ethyleneglycolether) 9016-45-9 ( <0.9 ), Polyethylene glycol 25322-68-3 ( 7-13 ), Sucrose 57-50-1 ( 10-30 )
Includes • 10X DreamTaq Green Buffer (with 20 mM MgCl2), 4 x 1.25 mL

Store in a non-frost-free freezer at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.