Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X)
Thermo Scientific DreamTaq Green PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq Green buffer, MgCl2, and dNTPs.
£114.00 - £453.00
Specifications
GC-Rich PCR Performance | Low |
---|---|
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Shipping Condition | Dry Ice |
For Use With (Application) | Standard PCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11813933
|
Thermo Scientific™
K1081 |
200 Reactions |
£114.00
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11816843
|
Thermo Scientific™
K1082 |
1000 Reactions |
£453.00
1000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific DreamTaq Green PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq Green buffer, MgCl2, and dNTPs. The master mix retains all features of DreamTaq DNA Polymerase. It is capable of robust amplification of up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.
Highlights
- Direct loading of PCR product on gel
- High yields and high sensitivity of PCR
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Robust amplification with minimal optimization
- Incorporates modified nucleotides, but does not incorporate dUTP (Cat. No. R0133)
Applications
- Routine PCR amplification of DNA fragments up to 6 kb
- Genotyping
- High throughput PCR
Specifications
Low | |
Standard | |
Standard PCR | |
1X | |
3'-A |
DreamTaq DNA Polymerase | |
Dry Ice | |
2X | |
No | |
SuperMix or Master Mix |
For Research Use Only. Not for use in diagnostic procedures.