Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
Brand: Thermo Scientific™ K1081
Additional Details : Weight : 1.51950kg
Description
- DreamTaq DNA Pol,
- dNTPs,
- Green buffer, density reagent and dyes for direct loading,
- supplied with Water,
- Nuclease free
Specifications
4 x 1.25 mL | |
Standard PCR, RT-PCR, Genotyping, Cloning | |
2X | |
200 |