Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
Brand: Thermo Scientific™ K1082
This item is not returnable.
View return policy
Description
- DreamTaq DNA Pol,
- dNTPs,
- Green buffer, density reagent and dyes for direct loading,
- supplied with Water,
- Nuclease free
Specifications
20 x 1.25mL | |
Standard PCR, RT-PCR, Genotyping, Cloning | |
2X | |
1000 |