missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase

Product Code. 15666154 Shop All Thermo Scientific Products
Click to view available options
Color:
Colorless
Green
Quantity:
200 U
2500 U
4 x 2500 U
500 U
Unit Size:
200 units
2500 units
2500µL
500 units
Each
Pack of 2500
Pack of 4
This item is not returnable. View return policy

Product Code. 15666154

Brand: Thermo Scientific™ EP1713

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

Thermo Scientific DreamTaq Hot Start DNA Polymerase, available in colorless and green formats, is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Buffer containing magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available with pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.

The 10X DreamTaq Green Buffer also includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Direct loading of PCR products on a gel with DreamTaq Green Buffer

Applications

  • Generation of PCR products for TA cloning
  • Routine PCR
  • Colony PCR
  • Genotyping
  • RT-PCR

Recommended Storage

Store at -20C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • DreamTaq Hot Start DNA Polymerase (5 U/μL), 5 x 100 μL
• 10X DreamTaq Green Buffer (with 20 mM MgCl2), 10 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Quantity 2500 U
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Hot Start Built-In Hot Start
Overhang 3'-A
Reaction Format Separate Components
Color Green
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.